Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900882

Sigma-Aldrich

Anti-NLRC5 (N-terminus) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CLR16.1, NLRC5, NOD-like receptor C5, NOD27, NOD4, Protein NLRC5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunoblotting: 1-2 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRC5(84166)

Immunogen

An 18 amino acid peptide near the amino terminus of human NOD4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle G Rodino et al.
Infection and immunity, 87(3) (2018-12-19)
Orientia tsutsugamushi is an obligate intracellular bacterium that infects mononuclear and endothelial cells to cause the emerging global health threat scrub typhus. The ability of O. tsutsugamushi to survive in monocytes facilitates bacterial dissemination to endothelial cells, which can subsequently
Muzi Wang et al.
Journal of cellular physiology, 234(11), 19990-20001 (2019-04-05)
NLRC5, the largest member of the Nod-like receptor (NLR) family, has been reported to play a pivotal role in regulating inflammatory responses. Recent evidence suggests that NLRC5 participates in Toll-like receptor (TLR) signaling pathways and negatively modulates nuclear factor-κB (NF-κB)
Ya-Ru Liu et al.
Journal of cellular physiology, 234(8), 14270-14284 (2019-01-16)
Rheumatoid arthritis (RA) is one of the chronic systemic autoimmune diseases that cardinally affect the joints. Many people all over the world suffer from the disease. Fibroblast-like synoviocytes (FLSs) play a significant role in the occurrence and development of RA.
Huisheng Liu et al.
Journal of virology, 93(11) (2019-03-22)
The role of nucleotide-binding oligomerization domain 2 (NOD2) in foot-and-mouth disease virus (FMDV)-infected cells remains unknown. Here, we showed that FMDV infection activated NOD2-mediated beta interferon (IFN-β) and nuclear factor-κB (NF-ĸB) signaling pathways. NOD2 inhibited FMDV replication in the infected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service