Skip to Content
Merck
All Photos(1)

Documents

S8701

Sigma-Aldrich

SFLLR-NH2 trifluoroacetate salt

≥95% (HPLC), powder

Synonym(s):

L-Seryl-L-phenylalanyl-L-leucyl-L-leucyl-L-Arginine amide trifluoroacetate salt, Ser-Phe-Leu-Leu-Arg-amide trifluoroacetate salt, Thrombin Receptor Activator for Peptide 5 (TRAP-5) trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H51N9O6 · xC2HF3O2
CAS Number:
Molecular Weight:
633.78 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥2 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CO)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O

InChI

1S/C30H51N9O6.C2HF3O2/c1-17(2)13-22(27(43)36-21(25(32)41)11-8-12-35-30(33)34)38-28(44)23(14-18(3)4)39-29(45)24(37-26(42)20(31)16-40)15-19-9-6-5-7-10-19;3-2(4,5)1(6)7/h5-7,9-10,17-18,20-24,40H,8,11-16,31H2,1-4H3,(H2,32,41)(H,36,43)(H,37,42)(H,38,44)(H,39,45)(H4,33,34,35);(H,6,7)/t20-,21-,22-,23-,24-;/m0./s1

InChI key

UUNMFWMQJXBTAA-VHIJBDTGSA-N

Application

SFLLR-NH2 trifluoroacetate salt may be used in cell signaling and immunology studies.

Biochem/physiol Actions

SFLLR-NH2 is a PAR-1 (Protease-Activated Receptor) selective activating peptide, also known as Thrombin Receptor Activating Peptide 5 (TRAP-5). Protease-activated receptors (PARs) are present on various organs including, platelets, mast cell, gallblader, oesophagus etc, and regulate various physiological processes including human platelet aggregation, vascular contraction/relaxation, and an increase in endothelial permeability. Recent papers indicated that PAR′s are also involved in sensory processing and proliferation of astrocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service