Skip to Content
Merck

S4522

Glass spreaders

90° bend

Synonym(s):

cell spreader, glass spreader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.14
UNSPSC Code:
41122103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


General description

Constructed of 4 mm diameter glass rod with polished ends, with 130 mm long handles and 50 mm long spreader segments.

Application

For inoculating petri dishes


Still not finding the right product?

Explore all of our products under Glass spreaders




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library