Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352202
MDL number:
recombinant
expressed in HEK 293 cells
UniProt accession no.
storage temp.
−70°C
Gene Information
rat ... Htr7(65032)
Packaging
Actual concentrations and specific binding capacity are provided with each lot.
Preparation Note
Membranes are suspended in a mixture containing 10 mM Tris-HCl at pH 7.2, 2 mM EDTA.
Analysis Note
Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.