RAB1754
Human Tryptase beta-2 ELISA
for serum, plasma and cell culture supernatants
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 ng/mL
standard curve range: 2.048-500 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TPSB2(64499)
Related Categories
General description
Tryptase β2 (TPSB2) gene codes for mast cell tryptase β2. It is expressed in whole blood by mast cells and their progenitors. Mast cell βII-tryptase protein is a S1A protease.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Biochem/physiol Actions
Tryptase β2 (TPSB2) can break down the large fibronectin substrate into multiple, smaller peptides ranging in size from approximately 30 to 50 kDa. It can also cleave fibrinogen. TPSB2 is associated with pathological inflammation.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S A Brooks et al.
Animal genetics, 41 Suppl 2, 16-22 (2010-11-26)
Inflammatory damage to the digital laminae, a structure responsible for suspension of the distal skeleton within the hoof capsule, results in a painful and often life-threatening disease in horses called laminitis. There can be many diverse causes of laminitis; however
Kristina M Cook et al.
The Journal of biological chemistry, 288(48), 34920-34929 (2013-10-22)
The S1A serine proteases function in many key biological processes such as development, immunity, and blood coagulation. S1A proteases contain a highly conserved disulfide bond (Cys(191)-Cys(220) in chymotrypsin numbering) that links two β-loop structures that define the rim of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service