RAB0267
Human IL-18 BPa ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 20 pg/mL
standard curve range: 24.69-18000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL18BP(10068)
Related Categories
General description
The Human IL-18 BPa ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-18 BPa in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human IL-18 BP α
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Benjamin Uzan et al.
EMBO molecular medicine, 6(6), 821-834 (2014-04-30)
Development of novel therapies is critical for T-cell acute leukaemia (T-ALL). Here, we investigated the effect of inhibiting the MAPK/MEK/ERK pathway on T-ALL cell growth. Unexpectedly, MEK inhibitors (MEKi) enhanced growth of 70% of human T-ALL cell samples cultured on
Zufeng Ding et al.
Biochemical and biophysical research communications, 451(4), 637-643 (2014-08-19)
Lectin-like oxidized low-density lipoprotein scavenger receptor-1 (LOX-1) is known to be involved in many pathophysiological events, such as inflammation. To clarify the role of LOX-1 in mtDNA damage and NLRP3 inflammasome activation, we studied wild-type (WT) and LOX-1 knockout (KO)
Peng Wang et al.
Molecular medicine reports, 24(1) (2021-05-27)
Heart failure (HF) is a progressive myocardial disease that affects pulse rate. Notably, chronic inflammation serves a crucial role in cardiac dysfunction and HF. Appropriate cardiomyocyte‑fibroblast communication is essential for cardiac function. In addition, cardiac fibroblasts (CFs) are the main
Kaori Sadakane et al.
Journal of applied toxicology : JAT, 34(8), 893-902 (2013-08-29)
Phthalate esters in plastics act as adjuvants for immunoglobulin production, which aggravates allergic disease. However, the effects of alkylphenols (used as plasticizers and surfactants) on atopic dermatitis have not been studied in detail. Therefore, the goal of the present study
J Michelle Kahlenberg et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(1), 152-162 (2014-01-23)
Systemic lupus erythematosus (SLE) is a systemic autoimmune syndrome associated with organ damage and an elevated risk of cardiovascular disease resulting from activation of both innate and adaptive immune pathways. Recently, increased activation of the inflammasome machinery in SLE has
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service