Skip to Content
Merck

RAB0011

Phospho-Akt (pSer473) ELISA Kit

for detection of human, mouse or rat phospho-akt (pSer473) in cell and tissue lysates

Synonym(s):

Phospho-Akt ELISA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... AKT1(207)
Input:
sample type cell lysate
sample type tissue lysate
Species reactivity:
human, rat, mouse
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human, rat, mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AKT1(207)

General description

The Phospho-Akt (pSer473) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

Akt (pSer473) synthetic phosphopeptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library