Skip to Content
Merck

PLA0113

Rabbit anti-Estrogen Receptor Alpha Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CTERP-1, ER, ER-alpha, ERalphaDup5, ERalphai45a, ERalphai45bL, ERalphai45bS, ERalphai45c, ERalphai56, ERalphai67, ESR, ESRA, ESTRR, Era, NR3A1, estradiol receptor, estrogen nuclear receptor alpha, estrogen receptor alpha E1-E2-1-2, estrogen receptor alpha E1-N2-E2-1-2, estrogen receptor alpha splice variant, nuclear receptor subfamily 3 group A member 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000

accession no.

P03372

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

Immunogen

The epitope recognized by PLA0113 maps to a region between residue 550 and the C-terminus (residue 595) of human Estrogen Receptor Alpha using the numbering given in SwissProt entry P03372 (GeneID 2099).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

The estrogen receptor (ER) is the steroid-hormone nuclear receptor that binds estrogen and functions to activate genes involved in a variety of biological processes. Two forms of the ER, ER alpha and ER beta, are encoded by independent genes, and each plays different roles in gene regulation. Additional variants of ER beta have also been reported; these include ERb2, ERb3, ERb4, and ERb5. All forms of the ER have the ability to heterodimerize with each other. Differences in localization and concentrations as well as heterodimerization among the variants may serve to achieve distinct cell and tissue responses. ER alpha generally functions as an activator of transcription while ER beta functions as a repressor.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library