Skip to Content
Merck

Skip To

P4560

Phosphate-Citrate Buffer with Urea Hydrogen Peroxide

tablet

Synonym(s):

P-C Buffer with Urea Hydrogen Peroxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tablet

Quality Segment

reaction suitability

reagent type: oxidant

solubility

water: 1 tablet/10 mL, clear, colorless

storage temp.

2-8°C

Application

Phosphate-Citrate Buffer Tablets with Urea Hydrogen Peroxide may be used as a peroxidase substrate buffer for ELISA with the H2O2 included.

Preparation Note

One tablet dissolved in 10 mL of deionized water yields a 0.05 M phosphate-citrate buffer, containing 0.014% H2O2, pH 5.0 at 25 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P9187HX0636HX0640
description

tablet

description

tablet

description

OmniTrace Ultra

description

liquid

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

tablet

form

tablet

form

liquid

form

liquid

solubility

water: 1 tablet/10 mL, clear, colorless

solubility

water: 0.5 tablet/10 mL, clear, colorless

solubility

soluble (Water: Soluble)

solubility

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

no temp limit

storage temp.

no temp limit

reaction suitability

reagent type: oxidant

reaction suitability

-

reaction suitability

-

reaction suitability

-

ACS grade dimethyl (2,2,2-trichloro-1-phenylsulfonylamino)ethylphosphonate

-

-

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the composition of the product p4560?

    1 answer
    1. This product contains citric acid, sodium phosphate, urea hydrogen peroxide, and other proprietary components, such that one tablet will yield 0.05 M phosphate-citrate buffer, containing 0.014% H2O2 when dissolved in 10 mL of deionized water.

      Helpful?

Reviews

No rating value

Active Filters