MTCP1001QPS
MTC™ Bio Pure.Amp™ qPCR Sealing Film
to fit standard qPCR plates, pressure adhesive
Synonym(s):
qPCR sealing membrane, sealing membrane, snap closure tube
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.15
Recommended Products
packaging
bag of 10 × 10 ea
manufacturer/tradename
MTC™ Bio P1001-QPS
Related Categories
General description
Sealing film, qPCR Optical (pressure-sensitive adhesive) ABI Type, 10 bags of 10 seals, 100/pk
Features and Benefits
- RNase, DNase, DNA, and PCR inhibitor free
- Evaporation rates ≤ 5%
- Inner packs of 10 membranes
- -40° to 120°C
- RNase, DNase, DNA, and PCR inhibitor free
- Inner packs of 10 membranes
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio