Skip to Content
Merck

MTCP1001E

MTC Bio Pure.Amp ELISA Plate Sealing Film

transparent, non-sterile

Synonym(s):

ELISA plate seal, cell sieve adaptor, flow cytometry tube, low adhesive membrane, plastic tube, sealing membrane, MTC Bio P1001-E, qPCR plate sealing membrance

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41122410
Manufacturer/tradename:
MTC Bio P1001-E
Sterility:
non-sterile
Packaging:
pack of × 100 ea (10 packs of 10ea)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

packaging

pack of × 100 ea (10 packs of 10ea)

manufacturer/tradename

MTC Bio P1001-E

color

transparent

General description

PureAmp Sealing Membranes compatible with all standard PCR plates for use in ELISA, EIA, and General Applications. Transparent film with low-adhesive EZ-peel, guaranteed to keep evaporation rates below 5%.

Features and Benefits

  • Certified RNase, DNase, DNA, and PCR inhibitor free
  • Evaporation rates <5%
  • Packed as 10 bags of 10 membranes
  • Temperature range -40° to 120°C

Legal Information

MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library