Skip to Content
Merck

MAK552

Fluo-4 No Wash Calcium Assay Kit

sufficient for fluorometric test plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116104
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; genitourinary disorders; neurological disorders; cancer

storage temp.

−20°C

General description

The Fluo-4 No Wash Calcium Assay Kit provides a homogeneous fluorescence-based assay for detecting the intracellular calcium mobilization. Cells expressing a GPCR of interest that signals through calcium are preloaded with Fluo-4 AM which can cross the cell membrane. Once inside the cell, the lipophilic blocking groups of Fluo-4 AM are cleaved by non-specific cell esterase, resulting in a negatively charged Fluo-4 dye that stays inside cells, and its fluorescence is greatly enhanced upon binding to calcium. When cells are stimulated with screening compounds, the receptor signals the release of intracellular calcium, which greatly increases the fluorescence of Fluo-4.


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library