Skip to Content
Merck

MAK262

Sphingomyelin Quantification Colorimetric Assay Kit

sufficient for 100 colorimetric tests

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


detection method

colorimetric

relevant disease(s)

neurological disorders

storage temp.

−20°C

General description

Sphingomyelin, a sphingolipid consisting of a phospholipid (typically phosphocholine) attached to a ceramide moiety, is a major component of cellular membranes. Sphingomyelin is enriched in the exoplasmic leaflet of the cell membrane. Within the cell membrane, sphingomyelin interacts with cholesterol to form lipid rafts. The metabolism of sphingomyelin contributes to cellular signaling via the release of ceramide, a second messenger that participates in multiple cellular signaling pathways. Sphingomyelin accumulates in Niemann-Pick disease due to deficiencies in sphingomyelinase activity. Increased levels of sphingomyelin is observed in hypercholesterolemia, which is characterized by the accumulation of sphingomyelin in arteriosclerotic lesions resulting in ischemic heart disease.

Application

Suitable for quantifying sphingomyelin in serum, plasma, and other biological fluids, and tissue and cell culture samples.

Biochem/physiol Actions

The Sphingomyelin Quantification Assay Kit is a simple and sensitive method for quantifying sphingomyelin. The assay is based on the hydrolysis of sphingomyelin into ceramide and phosphocholine by sphingomyelinase. Alkaline phosphatase (ALP) dephosphorylates phosphocholine to choline producing a reaction that generates a colorimetric signal (A570). The assay is sensitive to 1 M of sphingomyelin in a variety of samples.


signalword

Danger

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library