Skip to Content

M8927

Malt Extract Agar, modified

BioChemika, suitable for microbiology

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41171605
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

pH

4.7±0.2 (25 °C)

application(s)

food and beverages, microbiology

Application

Recommended for isolation, detection and enumeration of yeasts and molds.

Other Notes

Components (g/L)
Peptic digest of animal tissue 0.78
Maltose 12.75
Dextrin 2.75
Agar 15.00


Still not finding the right product?

Explore all of our products under Malt Extract Agar, modified


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library