Merck
L8519
Sigma label tape
red
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.35
UNSPSC Code:
55121611
Manufacturer/tradename:
Sigma-Aldrich
Material:
red
Packaging:
pkg of 1 roll
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
red
packaging
pkg of 1 roll
manufacturer/tradename
Sigma-Aldrich
W
3/4 in. (1.91 cm)
core size
3 in. (7.62 cm)
roll size
2160 in. (55 m)
General description
All purpose, self-adhesive tape can be written on with pen, pencil or permanent marker. Temperatures from -23°C to +121°C will not affect tape.
Still not finding the right product?
Explore all of our products under Sigma label tape
Choose from one of the most recent versions:
Certificates of Analysis (COA)