Skip to Content
Merck

K2644

Kisspeptin-234 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

kisspeptin-10 peptide 234

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C63H78N18O13 · xC2HF3O2
CAS Number:
Molecular Weight:
1295.41 (free base basis)
NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

film

color

colorless to white

solubility

H2O: >1 mg/mL

storage temp.

−20°C

Biochem/physiol Actions

Kisspeptin-234 is a 10-amino acid peptide antagonist with potent neutral antagonist activity at GPR-54 and competes directly at the Kisspeptin-10 binding site.
Kisspeptin-234 is a 10-amino acid peptide antagonist with potent neutral antagonist activity at GPR-54 and competes directly at the Kisspeptin-10 binding site. Kisspeptin-234 is the first reported antagonist for the kisspeptin-1/GPR-54 signaling system.

Gonadotropin-releasing hormone (GnRH) modulates the hypothalamic-pituitary-gonadal axis, enabling the brain to control reproductive processes. Kisspeptin-1 is a peptide neurotransmitter released from afferent neurons onto GnRH neurons, where it binds to and activates to the orphan receptor GPR-54 to regulate release of GnRH. Regulation of kisspeptin/GPR-54 signaling presents an opportunity for reproductive therapeutics above the level of GnRH and gonadotropin secretion. Pharmacological and selective tools are needed to better understand kisspeptin-1/GPR-54 signaling. Analogs of kisspeptin-10, a 10-amino acid portion of kisspeptin-1, were designed to be full and neutral antagonists of GPR-54.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library