Skip to Content
Merck

HS8610E

Laboratory Notebook

100 pages, lined, black

Synonym(s):

experiment notebook, lab notebook

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
14111514
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

lined

packaging

pk of 12

manufacturer/tradename

Heathrow Scientific HS8610E

L × W

218 × 281 mm

color

black

General description

Use these notebooks to keep information safe and well organized. Record research data, experimental procedures, and design ideas.

Features and Benefits

  • numbered, acid-free, pages include spaces for title, project number, book number, date, and witness signature
  • lined notebooks feature college-ruled pages for easy report writing
  • covers for all versions are waterproof and chemical resistant
  • all notebooks contain periodic and conversion tables in the back



Still not finding the right product?

Explore all of our products under Laboratory Notebook




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library