Skip to Content
Merck
All Photos(4)

Key Documents

HPA005788

Sigma-Aldrich

Anti-GRAP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Adapter protein GRID antibody produced in rabbit, Anti-GADS protein antibody produced in rabbit, Anti-GRB-2-like protein antibody produced in rabbit, Anti-GRB2-related adapter protein 2 antibody produced in rabbit, Anti-GRB2L antibody produced in rabbit, Anti-GRBLG antibody produced in rabbit, Anti-GRBX antibody produced in rabbit, Anti-Grf-40 antibody produced in rabbit, Anti-Grf40 adapter protein antibody produced in rabbit, Anti-Growth factor receptor-binding protein antibody produced in rabbit, Anti-Hematopoietic cell-associated adapter protein GrpL antibody produced in rabbit, Anti-P38 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

TNSISRQKQIFLRDRTREDQGHRGNSLDRRSQGGPHLSGAVGEEIRPSMNRKLSDHPPTLPLQQHQHQPQPPQYAPAPQQLQQPPQQRYLQHHHFHQERRGGSLDINDGHCGTGLGSEMNAALMHRRHTDPVQLQAAGRVRWARALYDFE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRAP2(9402)

Immunogen

GRB2-related adapter protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GRB2-related adapter protein 2 is a protein encoded by the GRAP2 gene in humans. It is referred as P38, GADS, GRID, GRPL, GrbX, Mona, GRB2L, GRBLG, Grf40 and GRAP-2. It is involved in tyrosine kinase signaling in hematopoietic cells as well as plays a tissue-specific role as an inhibitor of RET mitogenic signaling. Its expression is restricted to hematopoietic cells and plays an important role in intracellular signaling in T cells, monocytic cells and platelets.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73396

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leopold Ludwig et al.
Oncogene, 22(34), 5362-5366 (2003-08-15)
Signal transduction of the RET receptor tyrosine kinase is involved in developmental processes as well as in neoplastic transformation. Activation of RET initiates receptor autophosphorylation on specific tyrosines that act as docking sites for downstream signaling molecules. Using the cytoplasmatic
B Guyot et al.
Gene, 290(1-2), 173-179 (2002-06-14)
Monocytic adaptor (Mona) also known as Gads is a Grb2-related adaptor whose expression is restricted to hematopoietic cells. It plays an important role in intracellular signaling in T cells, monocytic cells, and platelets. Here we investigated the regulatory aspects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service