Skip to Content
Merck
All Photos(1)

Key Documents

GW22382A

Sigma-Aldrich

Anti-G3BP antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

manufacturer/tradename

Genway 15-288-22382A

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G3BP1(10146)

Immunogen

Immunogen Sequence: GI # 5031703, sequence 132-350
Recombinant ras-GTPase-activating protein SH3-domain-binding protein

Application

Anti-G3BP antibody produced in chicken is suitbale for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Ras GTPase-activating protein-binding protein 1 is an enzyme encoded by the G3BP1 gene in humans. It is a ubiquitously expressed cytosolic 68kDa protein that physically associates with the SH3 domain of GAP and is essential for Ras signaling. The protein binds to RasGAP and is overexpressed in many human cancers. Down regulation of G3BP1 suppresses cell growth and induces apoptosis in HCT116 cells. It acts as a potential anti-cancer drug target implicated in several cellular functions.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhang et al.
Cancer gene therapy, 20(11), 622-629 (2013-10-26)
G3BP is a RasGAP binding protein that is overexpressed in many human cancers. We previously reported that downregulation of G3BP suppressed cell growth and induced apoptosis in HCT116 cells. Here we report that both transient and stable knockdown of G3BP
Tina Vognsen et al.
PloS one, 8(12), e80947-e80947 (2013-12-11)
Ras GTPase Activating Protein SH3 Domain Binding Protein (G3BP) is a potential anti-cancer drug target implicated in several cellular functions. We have used protein crystallography to solve crystal structures of the human G3BP1 NTF2-like domain both alone and in complex
F Parker et al.
Molecular and cellular biology, 16(6), 2561-2569 (1996-06-01)
We report the purification of a Ras-GTPase-activating protein (GAP)-binding protein, G3BP, a ubiquitously expressed cytosolic 68-kDa protein that coimmunoprecipitates with GAP. G3BP physically associates with the SH3 domain of GAP, which previously had been shown to be essential for Ras

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service