Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9374-52

DIGE Buffer Kit

Cytiva 28-9374-52, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.25
Pricing and availability is not currently available.

sterility

non-sterile

form

liquid

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 28-9374-52

storage temp.

2-8°C

General description

Dige buffer kit is a tool to increase the quality, reproducibility, and convenience of 2-d dige using ettan dige system. The dige buffer kit is a specially designed two-buffer system for running up to twelve dige gels simultaneously.

Dige gel should be used with the dige buffer kit, which consists of concentrated running buffers and sealing solution for attaching immobilinedrystrip gels (ipg strips) to the top of the polyacrylamide gel. the capacity of the buffer system is similar to the commonly used laemmli (tris-glycine) buffer system, and the separation performance of dige gel is comparable to 12.5% laemmli gels (1). dige gel and dige buffer kit offer the high quality and performance of the laemmli system with the added advantages of convenience and time savings.

Dige gel is intended for use with the dige buffer kit and ettan dalt large vertical system. ettan dalttwelve and ettan daltsix instruments are designed for large-format, second dimension 2-d electrophoresis using either 18 or 24 cm immobiline drystrip gels. with ettan dalttwelve, up to 12 dige gels can be run simultaneously with separations complete in 4 to 6 h. the dige buffer kit contains the amount of reagents necessary for a single run of up to 12 dige gels using the ettan dalttwelve electrophoresis unit, or two runs of up to six gels using the ettan daltsix unit.

Features and Benefits

  • Precast 12.5% polyacrylamide buffer for second-dimension 2-D electrophoresis.
  • Ready-to-use products with a long shelf-life that saves you time by eliminating the need to make buffers, cast gels, and wash glass plates, leaving more time for more important work.
  • DIGE gels run with DIGE Buffer Kit result in spot maps with higher reproducibility compared to homecast polyacrylamide gels.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

This page describes two-dimensional separation of protein samples with Cytiva products.

This page describes two-dimensional separation of protein samples with Cytiva products.

This page describes two-dimensional separation of protein samples with Cytiva products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service