GE17-0403-03
HiTrap® Protein A High Performance
Cytiva 17-0403-03, pack of 5 × 5 mL
Synonym(s):
HiTrap Protein A HP, high performance column, antibody purification
About This Item
Recommended Products
ligand
native protein A
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 17-0403-03
storage condition
(20% Ethanol)
parameter
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
size
avg. part. size
34 μm
cleaning
2-10
working range
3-9
storage temp.
2-8°C
General description
HiTrap® Protein A HP is a prepacked ready to use, column for preparative purification of monoclonal and polyclonal antibodies. The special design of the column, together with the medium provide fast, simple and easy separations in a convenient formulation.
Features and Benefits
- High binding capacitiy of 20 mg IgG/mL medium and fast kinetics.
- 34 μm bead size for high-performance, high-resolution purifications.
- Designed for use with syringe, pump, or chromatography system such as AKTA design or FPLC™ System.
- Stable over pH 3 to 9 for use, and 2 to 10 for cleaning.
- Robust, easy-to-use method.
Storage and Stability
Analysis Note
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page shows how to purify IgM antibodies by affinity chromatography using HiTrap™ IgM Purification HP.
This page shows how to separate IgG antibodies by affinity chromatography using HiTrap® Protein G HP columns from Cytiva.
Protocols
Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.
Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.
Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.
Protein A, coupled to Sepharose, binds IgG via its Fc regions, facilitating affinity chromatography purification.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service