Skip to Content
Merck
All Photos(1)

Key Documents

EP5420000040

Eppendorf® Centrifuge 5420

keypad, refrigerated: no, with rotor FA-24x2, AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug

Synonym(s):

Centrifuge 5420 - Microcentrifuge, Eppendorf® 120 V Centrifuge 5420

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

CE compliant
CSA approved
refrigerated: no
keypad
with rotor FA-24x2

Application

Centrifugation

Features and Benefits

with keypad control panel for easy cleaningwith aerosol-tight Rotor FA-24x2non-refrigerated120 V, 50 – 60 Hz

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service