Skip to Content
Merck
All Photos(1)

Key Documents

EP022491776

Eppendorf® epT.I.P.S.® Reloads, PCR clean

volume 0.25-2.5 mL, L 115 mm, red, colorless tip, pack of 480 ea (10 trays × 48 ea)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600

material

colorless tip
polypropylene
red

sterility

non-sterile

feature

DNase free
RNase free

packaging

pack of 480 ea (10 trays × 48 ea)

manufacturer/tradename

Eppendorf® 022491776

L

115 mm

volume

0.25-2.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

epT.I.P.S.® Reloads, PCR clean, 0.25 – 2.5 mL, 115 mm, red, colorless tips, 480 tips (10 trays × 48 tips). Replaces order no. 0030073886.
  • Reduced waste as compared with disposable racks
  • The refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
  • The refill system, depending on tips size, is packaged as either dual-sided or in stack form
  • Available in purity grades of Eppendorf Quality and PCR clean
  • Color-coded trays for simple volume and pipette identification

Features and Benefits

  • Reduced waste as compared with disposable racks
  • The refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
  • The refill system, depending on tip size, is packaged as either dual-sided or in stack form
  • Available in purity grades of Eppendorf Quality and PCR clean
  • Color-coded trays for simple volume and pipette identification

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service