Skip to Content
Merck
All Photos(1)

Key Documents

EHU113961

Sigma-Aldrich

MISSION® esiRNA

targeting human PARK7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTGAAGGAGCAGGAAAACCGGAAGGGCCTGATAGCCGCCATCTGTGCAGGTCCTACTGCTCTGTTGGCTCATGAAATAGGTTTTGGAAGTAAAGTTACAACACACCCTCTTGCTAAAGACAAAATGATGAATGGAGGTCATTACACCTACTCTGAGAATCGTGTGGAAAAAGACGGCCTGATTCTTACAAGCCGGGGGCCTGGGACCAGCTTCGAGTTTGCGCTTGCAATTGTTGAAGCCCTGAATGGCAAGGAGGTGGCGGCTCAAGTGAAGGCTCCACTTGTTCTTAAAGACTAGAGCAGCGAACTGCGACGATCACTTAGAGAAACAGGCCGTTAGGAATCCATTCTCACTGTGTTCGCTCTAAACAAAACAGTGGTAGGTTAATGTGTTCAGAAGTCGCTGTCCTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prahlad V Raninga et al.
Apoptosis : an international journal on programmed cell death, 21(12), 1422-1437 (2016-10-28)
Multiple myeloma (MM) is an incurable plasma B cell malignancy. Despite recent advancements in anti-MM therapies, development of drug resistance remains a major clinical hurdle. DJ-1, a Parkinson's disease-associated protein, is upregulated in many cancers and its knockdown suppresses tumor
Bijun Qiu et al.
Oncotarget, 8(5), 8499-8511 (2016-12-31)
Chronic liver inflammation and injuries play a critical role in development of hepatocellular carcinoma (HCC). Parkinson disease (autosomal recessive, early onset) 7, encoding PARK7 protein (also called DJ-1), plays important roles in many carcinogenesis processes and is essential in modulating
Hua-Zong Zeng et al.
International journal of molecular sciences, 12(6), 3489-3499 (2011-07-13)
The aim of study is to identify cisplatin-resistance associated biomarkers for non-small cell lung cancers (NSCLC). We use two-dimensional electrophoresis (2-DE) combined with MALDI-TOF mass spectrometry to compare the proteome between lung cancer cell line A549 and its cisplatin-resistant subline
Canan Schumann et al.
Molecular pharmaceutics, 13(6), 2070-2083 (2016-05-14)
We report an efficient therapeutic modality for platinum resistant ovarian cancer based on siRNA-mediated suppression of a multifunctional DJ-1 protein that is responsible for the proliferation, growth, invasion, oxidative stress, and overall survival of various cancers. The developed therapeutic strategy
Karim Bahmed et al.
American journal of physiology. Lung cellular and molecular physiology, 317(4), L475-L485 (2019-07-18)
The alveolus participates in gas exchange, which can be impaired by environmental factors and toxins. There is an increase in using electronic cigarettes (e-cigarettes); however, their effect on human primary alveolar epithelial cells is unknown. Human lungs were obtained from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service