Skip to Content
Merck

D6539

2-Deoxy-α-D-ribose 1-phosphate bis(cyclohexylammonium) salt

≥97% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H11O7P · 2C6H13N
CAS Number:
Molecular Weight:
412.46
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Beilstein/REAXYS Number:
3785872
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥97% (TLC)

form

powder

color

white

solubility

water: 50 g/L at 20 °C

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.OC[C@H]3O[C@@H](C[C@@H]3O)OP(O)(O)=O

InChI

1S/2C6H13N.C5H11O7P/c2*7-6-4-2-1-3-5-6;6-2-4-3(7)1-5(11-4)12-13(8,9)10/h2*6H,1-5,7H2;3-7H,1-2H2,(H2,8,9,10)/t;;3-,4+,5+/m..0/s1

InChI key

GVYUZZDUVZHJAM-CGKAESPRSA-N

Application

2-Deoxyribose 1-phosphate (2dR1P) may be used in pyrimidine metabolism research to identify and characterize thymidine phosphorylase(s)/platelet-derived endothelial cell growth factor(s)(PD-ECGF)/gliostatin(s). 2-Deoxyribose 1-phosphate may be used to study processes such as angiogenesis in tumor cells at the level of thymidine phosphorylase activity. 2-Deoxy-a-D-ribose 1-phosphate (dR-1-P) is used in the organic synthesis of 2′-deoxynucleosides such as cladribine.

Preparation Note

Enzymatically prepared

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library