CLS355001
Corning® Falcon® Cell Culture Flask
capacity 150 mL, canted neck, graduated, 25 ‑ 325 mL, cap, blue vented
Synonym(s):
Corning container
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
polystyrene
feature
cap blue vented
graduated 25 ‑ 325 mL
manufacturer/tradename
Corning 355001
technique(s)
cell culture | mammalian: suitable
capacity
150 mL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service