CLS353801
Corning® Primaria Cell Culture Dish
W × L 35 mm × 10 mm, easy grip style
Synonym(s):
plastic cell culture dish
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.14
Recommended Products
material
polystyrene
manufacturer/tradename
Corning 353801
technique(s)
cell culture | mammalian: suitable
W × L
35 mm × 10 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service