CLS3343
Corning® 96 Well Storage Microplate
V-bottom, expanded volume, polypropylene, standard height, bag of 10 ×, non-sterile, lid: no
Synonym(s):
96 multiwell plate
About This Item
polypropylene
Recommended Products
material
V-bottom
polypropylene
sterility
non-sterile
feature
lid: no
manufacturer/tradename
Corning 3343
packaging
bag of 10 ×
maximum volume
450 μL
size
96 wells
working volume
450 μL
color
clear
suitability
suitable for (general storage, liquid handling; automated applications)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Corning® 96 well microplates support various assay and storage application requirements.
- Features uniform skirt heights for the greater robotic gripping surface.
- Resistant to most solvents including DMSO, ethanol, methanol.
- RNase-/DNase-free.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service