Skip to Content
Merck
All Photos(1)

Key Documents

MES-SA/Dx-5

95051031, human uterus, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

Product Name

MES-SA/Dx-5, 95051031

biological source

human uterus

growth mode

Adherent

karyotype

Modal no. 47 (parent line MES-SA-45). Reported karyotype 47,XX,12qt,22q-t(5q,6p),-3,-4,-7,+t(3q?,7q),Mar1,+mar2,+Mar3

morphology

Epithelial

products

High levels of Mdr1 mRNA and P-glycoprotein

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human uterus sarcoma

Cell Line Description

The multi drug-resistant cell line MES-SA/Dx5 was derived from the human uterine sarcoma cell line MES-SA (Sigma Catalogue number. 95051030) which was originally obtained from a tumour from a 56-year-old Caucasian female at the time of hysterectomy. The Dx5 variant exhibits a 100-fold resistance to doxorubicin and has a reported doubling time of 30 hours. The two additional marker chromosomes indicate clonal selection during drug selection. MES-SA/Dx-5 cells exhibit marked cross-resistance to a number of chemotherapeutic agents (including daunorubicin, dactinomycin, vincristine, taxol, colchicine) and moderate cross-resistance to mitomycin C and melphalan. Cross resistance to bleomycin, cisplatin, carmustine, 5-fluorouracil or methotrexate was not observed.

Application

Tumour toxicity assays and multi-drug-resistant studies.

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 to 1:8 i.e. seeding at 1-3 x 10,000 cells/cm2 using 0.03% EDTA; 5% CO2; 37°C. Tap side of flask to dislodge cells.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service