Skip to Content
Merck
All Photos(1)

Key Documents

BRL-1

94121649, rat liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
rat liver
growth mode:
Adherent
karyotype:
Diploid or pseudodiploid
morphology:
Epithelial
products:
Not specified
receptors:
Not specified

Product Name

BRL-1, 94121649

biological source

rat liver

growth mode

Adherent

karyotype

Diploid or pseudodiploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat epithelial cells

Cell Line Description

Isolated by Dr Hayden Coon. Flat epithelial cell colonies that appear upon low density plating as suspensions of collagenase dissociated buffalo rat liver. Cell origin not clearly defined.

Application

Feeder cells for co-culture with freshly isolated hepatocytes.

Culture Medium

Coon′s modified Ham′s F12 + 2mM Glutamine + 5% Foetal Bovine Serum (FBS) or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol + 5% Foetal Bovine Serum (FBS) + 2mM Glutamine.

Subculture Routine

Spit confluent cultures 1:100 i.e. seeding at 1x1,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service