Skip to Content
Merck
All Photos(1)

Documents

DH82

94062922, dog blood, Macrophage

Synonym(s):

DH82 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DH82, 94062922

biological source

dog blood

growth mode

Suspension

karyotype

Not specified

morphology

Macrophage

products

Not specified

receptors

FC-gamma receptors

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Canine Monocyte-macrophage

Cell Line Description

Derived from a ten year old male golden retriever with malignant histiocytosis. The cells have a macrophage-like morphology and are able to phagocytose latex particles. They are positive for Fc-gamma receptors and are negative for Fc-mu and C3b receptors. DH82 cells do not produce IL-1.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Maintain cultures at 3-9x100,000 cells/ml; 5% CO2; 37°C. Cells are prone to sticking and can be removed with 0.02% EDTA.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service