Skip to Content
Merck
All Photos(1)

Key Documents

Sf21

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

insect ovary

packaging

tube of 5 μg 05022801-DNA-5UG
pkg of vial of cells 05022801-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Spherical

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Spodoptera frugiperda pupal ovarian tissue

Cell Line Description

Derived from pupal ovarian tissue of spodoptera frugiperda. The cells are highly susceptible to Baculovirus infection and are used in the production of protein products genetically manipulated into Baculovirus vector systems. They were derived at the USDA Insect Pathology Laboratory from a cell line originating from pupal ovarian tissue of the fall army worm, Spodoptera frugiperda. Sf21 cells have greater range in size and have a doubling time of 24 hr compared to 48-72 hr for Sf9 cells.

Application

Protein expression using the Baculovirus vector system

Culture Medium

SF90011 + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 5x10,000 cells/cm2; 28 C. Gently resuspend cells in old culture medium by dislodging the cells by pipetting, or gentle agitation, centrifuge suspension and resuspend cell pellet in fresh medium.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service