C6317
Nalgene® rectangular carboy
nominal capacity 9 L (2 gal), Style 2211, autoclavable: no
Synonym(s):
carboys, nalgene carboys
About This Item
material
high-density polyethylene Carboy
polypropylene screw closure
feature
autoclavable: no
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 2211-0020
L × W × H
8 1/2 in. (216 mm) × 5 3/4 in. (146 mm) × 13 1/2 in. (343 mm)
nominal capacity
9 L (2 gal)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service