Skip to Content
Merck

C-39115

Mammary Epithelial Cell Growth Medium

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-21210

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Mammary Epithelial Cell Growth Medium SupplementMix (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-21210

Application

Mammary Epithelial Cell Growth Medium is a serum-free medium developed for the cultivation of epithelial cells from the mammary gland. The medium is optimized for primary human cells but can also be used for rat and murine mammary epithelial cells as well as mamma carcinoma cell lines.Recommended for:Human Mammary Epithelial Cells

Preparation Note

Click here for more information.
Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 10 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.5 µg / ml
Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Analysis Note

All lots of PromoCell Mammary Epithelial Cell Growth Medium are subjected to comprehensive quality control tests using primary human mammary epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested mammary epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Other Notes

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library