Skip to Content

C2312

Nalgene® Labtop cooler

−20°C Labtop cooler, gel-filled lid

Synonym(s):

PCR amplification cooler, PCR amplification workstation, Nalgene 5115-0032, cryogenic storage, vial cryo storage box

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41103025
Manufacturer/tradename:
Nalgene® 5115-0032
Feature:
for tubes 0.2 ‑ 0.5 mL
Packaging:
pack of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


feature

for tubes 0.2 ‑ 0.5 mL

packaging

pack of 1 ea

manufacturer/tradename

Nalgene® 5115-0032

L × W × H

243 mm × 157 mm × 146 mm

General description

Maintains 32 microtubes below −15°C for up to 2 hrs at room temperature.
Molded from durable polycarbonate and filled with non-toxic freezable gel. A convenient alternative to ice buckets for keeping reagents and enzymes cool. Also protects critical samples from temperature fluctuations in the freezer.

Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nalgene® Labtop cooler




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library