Skip to Content
Merck

B020002

Sigma Black 2

Synonym(s):

Phosphoramidous acid, bis(1-methylethyl)-, 2-[[2-[bis(4-methoxyphenyl)phenylmethoxy]ethyl][4-[[2,5-dimethoxy-4-[(4-nitrophenyl)azo]phenyl]azo]phenyl]amino]ethyl 2-cyanoethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C54H61N8O9P
CAS Number:
Molecular Weight:
997.08
UNSPSC Code:
41116105
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

-10 to -25°C

Quality Level





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B020002-0.25G04065269804864
B020002-0.1MMOL04065269804857