Skip to Content
Merck

AXYBF400S

Breathable Rayon sealing film, for 50 μm deep well tissue culture plates

size 96 wells, pkg of 10x50sheets/cs, sterile

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122410
Manufacturer/tradename:
Axygen Inc BF-400-S, Corning
Sterility:
sterile
Packaging:
pkg of 10x50sheets/cs
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile

form

film

manufacturer/tradename

Axygen Inc BF-400-S, Corning

packaging

pkg of 10x50sheets/cs

size

96 wells

General description

  • Microplate sealing films are ideal for sealing plates in various laboratory applications including PCR and Real-Time PCR to ELISA and cell culture
  • The breathable rayon sealing films prevent sample contamination and are also suited for storage applications
  • Chemically resistant and can withstand varying temperatures
  • Enables efficient gas exchange for the cultivation of cellular and bacterial cultures
  • Suited for sealing polypropylene/polystyrene culture microplates, 96-well/384-well microplates, and other assay microplates


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library