Skip to Content
Merck
All Photos(2)

Key Documents

AV40421

Sigma-Aldrich

Anti-PTBP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HNRNPI, Anti-HNRPI, Anti-MGC10830, Anti-MGC8461, Anti-PTB, Anti-PTB-1, Anti-PTB-T, Anti-Polypyrimidine tract binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
rat, human, horse, guinea pig, mouse, bovine
citations:
1
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

58 kDa

species reactivity

rat, human, horse, guinea pig, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTBP1(5725)

General description

Polypyrimidine tract-binding protein 1 (PTBP1) is a member of the ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs) subfamily. It has an N-terminal nuclear shuttling domain and four repeats of quasi-RNA recognition motif (RRM) domains that bind RNAs. PTBP1 is mostly expressed in all cell types. The PTBP1 gene is mapped on human chromosome 19p13.3.

Immunogen

Synthetic peptide directed towards the N terminal region of human PTBP1

Biochem/physiol Actions

Polypyrimidine tract-binding protein 1 (PTBP1) plays a key role in T cell activation, spermatogenesis, and splicing. It is involved in the differentiation and development of neuronal cells, growth of embryos and erythrocytes. PTBP1 also participates in apoptosis, glycolysis, migration, metastasis, proliferation, and carcinogenesis due to its role in cancer as a splicing factor. This protein plays a major role in neurodegenerative diseases, cardiovascular diseases, colon cancer, colorectal cancer, breast cancer, glioma, and glioblastoma.
The hnRNPs are RNA-binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. PTBP1 protein binds to the intronic polypyrimidine tracts that requires pre-mRNA splicing and acts via the protein degradation ubiquitin-proteasome pathway. It may also promote the binding of U2 snRNP to pre-mRNAs. Alternatively spliced transcript variants encoding different isoforms have been described.

Sequence

Synthetic peptide located within the following region: RGSDELFSTCVTNGPFIMSSNSASAANGNDSKKFKGDSRSAGVPSRVIHI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhu et al.
Journal of Zhejiang University. Science. B, 21(2), 122-136 (2020-03-03)
Polypyrimidine tract-binding protein 1 (PTBP1) plays an essential role in splicing and is expressed in almost all cell types in humans, unlike the other proteins of the PTBP family. PTBP1 mediates several cellular processes in certain types of cells, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service