Skip to Content
Merck

A211850

DMT-2′O-MOE-rA(bz) Phosphoramidite

Synonym(s):

5′-Dimethoxytrityl-N6-Benzoyl-3′-[(2-cyanoethyl)- (N,N-diisopropyl)]-phosphoramidite- 2′-O-(2-methoxyethyl)adenosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C50H58N7O9P
CAS Number:
Molecular Weight:
932.01
MDL number:
NACRES:
NA.51
UNSPSC Code:
41116105
Assay:
≥98.0% (reversed phase HPLC)
Biological source:
non-animal source
Form:
powder
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

assay

≥98.0% (reversed phase HPLC)

form

powder

λ

conforms (UV/VIS Identity)

storage temp.

−20°C

General description

DMT-2′O-MOE-rG(ib) Phosphoramidite is a phosphoramidite that belongs to the family of amides of trivalent phosphorus acid H3PO3. It is an adenosine analog.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library