Skip to Content
Merck

Skip To

A1750

Acetobromo-α-D-glucose

≥95% (TLC)

Synonym(s):

1-Bromo-α-D-glucose tetraacetate, 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H19BrO9
CAS Number:
Molecular Weight:
411.20
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
96669
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥95% (TLC)

form

powder

optical activity

[α]25/D 188 to 202 °, c = 1 in chloroform

contains

1% CaCO3 as stabilizer

color

white to off-white

mp

88.5 °C ((191.3 °F ))

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C14H19BrO9/c1-6(16)20-5-10-11(21-7(2)17)12(22-8(3)18)13(14(15)24-10)23-9(4)19/h10-14H,5H2,1-4H3/t10-,11-,12+,13-,14+/m1/s1

InChI key

CYAYKKUWALRRPA-RGDJUOJXSA-N

Application

Acetobromo-α-D-glucose is used as a possible poly (ethylene terephthalate) (PET) surface modification reagent to increase its blood compatibility.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7562A8292G2354
assay

≥95% (TLC)

assay

≥93% (TLC)

assay

≥93% (GC)

assay

99%

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

-

biological source

-

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

mp

88.5 °C ((191.3 °F ))

mp

-

mp

80-110 °C (lit.)

mp

109-111 °C (lit.)


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters