Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
211F1.1, monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (387M-14), vial of 0.5 mL concentrate (387M-15), bottle of 1.0 mL predilute (387M-17), vial of 1.0 mL concentrate (387M-16), bottle of 7.0 mL predilute (387M-18)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100
isotype
IgG2aκ
control
astrocytoma
shipped in
wet ice
storage temp.
2-8°C
visualization
nuclear
Gene Information
human ... OLIG2(10215)
General description
Physical form
Preparation Note
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() NA |
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate - | conjugate unconjugated |
| clone 211F1.1, monoclonal | clone EP112, monoclonal | clone 211F1.1, monoclonal | clone polyclonal |
| antibody form culture supernatant | antibody form culture supernatant | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| species reactivity human | species reactivity human | species reactivity mouse | species reactivity mouse, rat, human |
| biological source mouse | biological source rabbit | biological source mouse | biological source rabbit |
| Gene Information human ... OLIG2(10215) | Gene Information human ... OLIG2(10215) | Gene Information human ... OLIG2(10215) | Gene Information human ... OLIG2(10215) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.







