Skip to Content
Merck
All Photos(1)

Key Documents

379R-1

Sigma-Aldrich

Carbonic Anhydrase IX (CA IX) (EP161) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP161, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (379R-17)
bottle of 7.0 mL predilute (379R-18)
vial of 0.1 mL concentrate (379R-14)
vial of 0.5 mL concentrate (379R-15)
vial of 1.0 mL concentrate (379R-16)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... CA9(768)

General description

Carbonic anhydrases are a family of zinc containing metalloproteins that catalyze the reversible hydration of CO<sub>2</sub>. Among these, carbonic anhydrase IX (CA IX) is anchored to the cell membrane and is expressed in the human gastrointestinal tract, chiefly in the stomach and gall bladder1. It is interesting to note that CA IX is overexpressed in epithelial malignancies of the uterus, cervix, lung, breast, and kidney; none of the associated normal tissues express this isozyme.1-4 CA IX is said to maintain the extracellular acidic pH, thus promoting cell growth in these tumors.5 Preliminary data suggest consistent immunoreactivity for anti-CA IX in clear cell renal cell carcinoma (RCC) with demonstrated sensitivity of 85% to 100%.2 Anti-CA IX, together with antibodies against PAX-2, Ksp-cadherin, and CD117, contributes to a robust panel that can be used to help make this distinction. Strong diffuse-to-multifocal immunostaining for anti-CA IX is observed in the large majority of urothelial carcinomas as opposed to the extremely weak and focal immunoreactivity seen in collecting duct carcinoma (CDC). Anti-CA IX can thus aid in distinguishing between urothelial carcinoma and CDC.6

Quality


IVD

IVD

IVD

RUO

Linkage

Carbonic Anhydrase IX (CA IX) Positive Control Slides, Product No. 379S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service