Skip to Content
Merck

24360C

EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined

Animal-component free, without L-glutamine, without hypoxanthine, without sodium bicarbonate, without thymidine, dry powder, suitable for cell culture

Synonym(s):

EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.71
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

EX-CELL® CD CHO, Serum-Free Medium for CHO cells, Chemically Defined Animal-component free, without L-glutamine, without hypoxanthine, without sodium bicarbonate, without thymidine, dry powder, suitable for cell culture. Use 19.24 g/L. See information sheet, 24360C @ www.sigmaaldrich.com for hydration instructions

Quality Segment

product line

EX-CELL®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

shipped in

ambient

storage temp.

2-8°C

General description

EX-CELL® CD CHO is a chemically defined, animal-component free, serum-free medium formulated without hypoxanthine, thymidine or L-glutamine, making it an appropriate medium for selection systems such as DHFR- and Glutamine Synthetase (GS System).

Application

EX-CELL® CD CHO is developed for thelong-term growth of Chinese Hamster Ovary (CHO) cells and expression ofantibodies or protein products in suspension cell culture. CHO suspensioncultures can be sub cultured directly into EX-CELL CD CHO fromserum-supplemented or serum-free medium with little or no adaptation.

Preparation Note

Formulated to contain 19.2 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library