Skip to Content
Merck

137258

MOPS sodium salt

EMPROVE® EVOLVE

Synonym(s):

MOPS sodium salt, 3-(N-Morpholino)propanesulfonic acid sodium salt, 4-Morpholinepropanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H14NNaO4S
CAS Number:
Molecular Weight:
231.25
UNSPSC Code:
12161700
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial; tested

pH

6.5-7.9

useful pH range

6.5-7.9

 

6.5

pKa (25 °C)

7.2

suitability

suitable for manufacturing use

storage temp.

2-30°C

SMILES string

[Na+].[O-]S(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S.Na/c9-13(10,11)7-1-2-8-3-5-12-6-4-8;/h1-7H2,(H,9,10,11);/q;+1/p-1

InChI key

MWEMXEWFLIDTSJ-UHFFFAOYSA-M

Application

MOPS Sodium is a biological buffer also referred to as a “Propane Sultone” buffer. The pKa of MOPS Sodium is 7.01 which make it an ideal candidate for medias and protein based buffer formulations to maintain a stable environment in solution. MOPS Sodium is considered to be non-toxic to culture cell lines and provides high solution clarity.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under MOPS sodium salt


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.