Skip to Content
Merck
All Photos(1)

Key Documents

Y0001249

Entacapone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Cyano-N,N-diethyl-3-(3,4-dihydroxy-5-nitrophenyl)propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N3O5
CAS Number:
Molecular Weight:
305.29
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

entacapone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1c(c(cc(c1)\C=C(\C(=O)N(CC)CC)/C#N)O)O

InChI

1S/C14H15N3O5/c1-3-16(4-2)14(20)10(8-15)5-9-6-11(17(21)22)13(19)12(18)7-9/h5-7,18-19H,3-4H2,1-2H3/b10-5+

InChI key

JRURYQJSLYLRLN-BJMVGYQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Entacapone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Entacapone is a catechol-O-methyl transferase (COMT) inhibitor. Used in treatment of Parkinson′s disease, entacapone is administered with L-DOPA to inihibit COMT from converting L-DOPA into a compound that cannot cross the blood brain barrier.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximiliano Rapanelli et al.
Behavioural brain research, 280, 92-100 (2014-12-02)
Dopamine encodes reward and its prediction in reinforcement learning. Catechol-O-methyltransferase (COMT) activity in the medial prefrontal cortex (mPFC) has been shown to influence cognitive abilities by modifying dopamine clearance. Nevertheless, it is unknown how COMT in the mPFC influences operant
Fabrizio Stocchi et al.
Parkinsonism & related disorders, 20(12), 1335-1340 (2014-10-13)
IPX066, an investigational extended-release carbidopa-levodopa (CD-LD) preparation, has demonstrated a rapid attainment and prolonged maintenance of therapeutic LD plasma concentrations in advanced Parkinson's disease (PD). This phase-3 crossover study assessed its efficacy and safety vs. CD-LD plus entacapone (CL + E). At
Paul E Polak et al.
Journal of neuroimmunology, 276(1-2), 135-141 (2014-09-23)
Reduced levels of noradrenaline (NA) in CNS of multiple sclerosis patients could be due to metabolism by catechol-O-methyltransferase (COMT). In mice immunized with myelin oligodendrocyte glycoprotein peptide, the BBB-permeable COMT inhibitor dinitrocatechol (DNC) reduced clinical signs, while entacapone, a non-BBB-permeable
Pasi Korhonen et al.
Movement disorders : official journal of the Movement Disorder Society, 30(5), 724-728 (2015-02-03)
The association between Parkinson's disease (PD) and prostate cancer, both common in elderly men, is disputable. In the STRIDE-PD study, prostate cancer developed in 9 patients (3.7%) receiving levodopa/carbidopa with entacapone, a catechol-O-methyltransferase inhibitor, versus 2 cases (0.9%) without entacapone.
Johanna Troberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 107-118 (2014-10-11)
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service