Skip to Content
Merck
All Photos(1)

Key Documents

TPO1000

Supelco

Total Phosphorus 1000 mg/L Calibration Standard

certified reference material

Synonym(s):

Phosphorus calibration solution (1000 mg/L)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC TPO1000-100

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I wish to know the chemical components used in the preparation of this P standard solution. TPO1000-100ML

    1 answer
    1. This item is made using two components: glycerol-2-phosphate and water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service