Skip to Content
Merck
All Photos(1)

Key Documents

PHR1560

Supelco

tert-Butyl Methyl Ether

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

tert-Butyl methyl ether, MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1083303

vapor density

3.1 (vs air)

CofA

current certificate can be downloaded

autoignition temp.

705 °F

expl. lim.

15.1 %

packaging

ampule of 3x1.2 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. tert-Butyl methyl ether (MTBE) is a gasoline additive. It is a commonly used organic solvent that can be synthesized by acid catalyzed reaction between methanol and isobutene. It is also an effective alternative to lead containing additives for enhancing the octane rating of gasoline.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2091 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceinwen A Schreiner et al.
Regulatory toxicology and pharmacology : RTP, 70(2 Suppl), S29-S34 (2014-05-24)
Micronucleus and sister chromatid exchange (SCE) tests were performed for vapor condensate of baseline gasoline (BGVC), or gasoline with oxygenates, methyl tert-butyl ether (G/MTBE), ethyl tert butyl ether (G/ETBE), t-amyl methyl ether (G/TAME), diisopropyl ether (G/DIPE), t-butyl alcohol (TBA), or
Yi-Hsueh Chuang et al.
Journal of hazardous materials, 283, 218-226 (2014-10-04)
During the chloramination of natural waters, both chloramines and dissolved organic nitrogen (DON) can serve as nitrogen sources for the formation of trichloronitromethane (TCNM) and dichloroacetonitrile (DCAN). The present study investigated the formation kinetics and precursor characteristics of TCNM and
Shilo Rosenwasser et al.
The Plant cell, 26(6), 2689-2707 (2014-06-13)
Marine viruses are major ecological and evolutionary drivers of microbial food webs regulating the fate of carbon in the ocean. We combined transcriptomic and metabolomic analyses to explore the cellular pathways mediating the interaction between the bloom-forming coccolithophore Emiliania huxleyi
Hiroki Toshima et al.
Journal of analytical toxicology, 39(2), 133-136 (2014-12-30)
During a cross-validation practice of urinary analysis of pyrethroid insecticide metabolite (3-phenoxybenzoic acid, 3-PBA) by two laboratories (Lab A and Lab B), difference in 3-PBA concentration was noticed. The analytical methods used in the exercise were enzymatic deconjugation (glucuronidase/sulfatase) followed
Jiaqi Liu et al.
Water research, 65, 64-72 (2014-08-05)
Using seawater for toilet flushing effectively reduces the consumption of precious freshwater resources, yet it introduces bromide and iodide ions into a wastewater treatment system, which may form bromo- and iodo-disinfection byproducts (DBPs) during chlorination of the wastewater effluent. Most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service