NIST953
Neutron density monitor wire
NIST® SRM® 953, cobalt in aluminum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
form
wire
packaging
pkg of 1 m
manufacturer/tradename
NIST®
General description
This Standard Reference Material (SRM) is provided as a reference source of cobalt in aluminum alloy to serve as a neutron density monitor wire standard. For more information, please refer ot the COA and SDS.
SRM 953_cert
SRM 953_SDS
SRM 953_cert
SRM 953_SDS
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Cobalt (Co)
See certificate for values and more details at nist.gov/SRM.
Cobalt (Co)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ni-Cr alloy as neutron flux monitor: composition and homogeneity assessment by NAA.
Fran?a E J, et al.
J. Radioanal. Nucl. Chem., 257(1), 113-115 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service