Skip to Content
Merck
All Photos(1)

Key Documents

NIST64C

Ferrochromium

NIST® SRM® 64c, high-carbon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This SRM is in the form of fine powder, less than 150μm (100 mesh) for use in checking chemical methods of analysis andin calibration with instrumental methods of analysis. For more information, please refer to the SDS and COA.

SRM 64C_cert

SRM 64C_SDS

Other Notes

Certified for the analytes listed below.
Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Iron (Fe), Manganese (Mn), Nickel (Ni), Nitrogen (N), Phosphorus (P), Silicon (Si), Sulfur (S), Titanium (Ti), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Arsenic (As), Molybdenum (Mo), Oxygen (O), Tin (Sn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Sol. 1 - Self-heat. 1 - Skin Sens. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service