Skip to Content
Merck
All Photos(2)

Key Documents

NIST57B

Silicon metal

NIST® SRM® 57b

Synonym(s):

Silicon

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 40 g

manufacturer/tradename

NIST®

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

application(s)

industrial qc

format

matrix material

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 57b contains a bottle consisting of approximately 40 grams of fine powder.



SRM 57B_cert

SRM 57B_SDS

Application

Silicon metal standard reference material (SRM) is intended for use in the evaluation of instrumental and chemical methods of analysis.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified values, expanded uncertainties, and expansion factor(k) for elements in SRM 57b.
  • Reference and information values are reported as mass fraction value for each element in silicon metal.
  • The certified values are metrologically traceable to the SI unit of mass, expressed as milligrams per kilogram.
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Boron (B), Phosphorus (P), Titanium (Ti), Manganese (Mn), Iron (Fe), Nickel (Ni), Zirconium (Zr)

Reference/Informational value is provided for the following analytes. Calcium (Ca), Cobalt (Co), Chromium (Cr), Copper (Cu), Vanadium (V), Hydrogen (H), Carbon (C), Oxygen (O), Sulfur (S)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon scaffold structured silicon anodes for lithium-ion batteries.
Guo J, et al.
Journal of Materials Chemistry, 20(24), 5035-5040 (2010)
High-Speed, Low-Power and Mid-IR Silicon Photonics Applications (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service